Would you like to log in again?
Quick Clean™ Safety Solvents and Degreasers, CRC
Supplier: CRC
Quick Clean™
125-03180
300050-438CS 193.64 USD InStock
300050-438
Quick Clean™ Safety Solvents and Degreasers, CRC
Cleaning Agents Detergents and Surface Cleansers
Fast evaporation, leaves no residue.
  • Non-flammable
  • Non-staining, non-corrosive, and non-conductive
  • High dielectric strength
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty