Would you like to log in again?
Dry Film PTFE Lubricants, LPS®
Supplier: LPS®
428-02616
300050-622CS 348.66 USD InStock
300050-622
Dry Film PTFE Lubricants, LPS®
Lubricants
  • Dry, thin film won't attract dust or dirt
  • Provides excellent lubrication
  • Ideal for releasing molded parts
  • Nonflammable
  • Does not contain silicone or chlorinated solvents
  • Safe on most surfaces
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty