Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Precision Clean Multi-Purpose Cleaner/Degreaser, LPS®

Supplier: LPS
428-02728
300050-626CS 238.23 USD InStock
300050-626
Precision Clean Multi-Purpose Cleaner/Degreaser, LPS®
Cleaning Agents Detergents and Surface Cleansers
Water-based biodegradable formula removes tough, stubborn stains and soils such as grease, oil, sludge, soot, ink, coffee, and more.

  • pH: 12.5–13.0
  • Does not contain chlorinated or petroleum solvents, bleach, ammonia, abrasives or butyl
  • Contains rust and corrosive inhibitor to prevent flash rusting of metal surfaces
  • Safe on all surfaces except magnesium
  • Nonflammable

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty