Would you like to log in again?
Silicone Lubricant, Loctite
Supplier: Loctite
442-51360
300051-108EA 46.98 USD InStock
300051-108
Silicone Lubricant, Loctite
Lubricants
For use with most metals, plastic or rubber.
  • Multi-purpose lubricant stops sticking, friction, squeaking, and binding of moving parts
  • Will not mar paint, rubber, or plastic surfaces
  • Waterproofs and electrically insulates
  • Non-gumming
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty