Would you like to log in again?
Industrial Quality All-Purpose Lubricant, L.S. Starrett
Supplier: L.S. Starrett
M1®
681-95173
300051-224CS 298.37 USD InStock
300051-224
Industrial Quality All-Purpose Lubricant, L.S. Starrett
Lubricants
  • M1 produces a micro-thin, airtight coating/film that simultaneously dries as it protects
  • M1 is free of silicone, making it an excellent lubricant
  • Deep-down penetration works quickly to free frozen nuts, bolts, and metal parts
  • Prevents rust
  • Actually removes grease, tar, and grime from metal parts and painted surfaces
  • Stops squeaks
  • Displaces moisture
  • Nonconductive
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty