Would you like to log in again?
Quick Connect Components, Western Enterprises
Supplier: Western Enterprises
312-QDB302 312-QDB300 312-QDB100 312-QDB101 312-QDB105 312-QDB301 312-QDB206 312-QDB200 312-QDB303 312-QDB201 312-QDB106 312-QDB205
300052-410EA 34.56 USD InStock
300052-410 300078-806 300052-412 300078-804 300078-812 300052-414 300078-802 300078-810 300078-800 300052-408 300078-798 300078-808
Quick Connect Components, Western Enterprises
Welding Supplies
  • Brass construction provides long service life
  • Cycle tested over 100,000 operations indicting a service life of 10 to 12 years of comparable use
  • Non-interchangeable oxygen and fuel gas connectors prevent accidental gas mixing
  • Instant closure on reverse flow check values with total shut-off at only 10 oz of back pressure
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty