Would you like to log in again?
Comfo Classic® Reusable Half Mask Respirators, MSA
Supplier: MSA
Comfo Classic®
454-808074 454-808073 454-808072 454-808071
300081-128EA 100.33 USD InStock
300081-128 300052-934 300052-936 300052-938
Comfo Classic® Reusable Half Mask Respirators, MSA
Respirators Reusable Respirators Half Face Respirators
SoftFeel® facepiece material increases the comfort factor dramatically.
  • Unique face seal design that gives wearers an exceptional fit
  • Facepiece made of silicone or Hycar™ rubber
Ordering information: Filter cartridge not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty