Would you like to log in again?
Sharpie® King Size Permanent Markers, ORS Nasco
Supplier: ORS Nasco
Sharpie®
586-15101PP
300053-086PK 34.38 USD InStock
300053-086
Sharpie® King Size Permanent Markers, ORS Nasco
Pens Permanent Markers
This is the solvent based (xylene) industrial strength permanent markers.
  • Durable aluminum barrel and felt tip stand up to heavy use
  • Large ink supply for big projects
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty