Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Rollgliss® Rope Rescue Systems, ORS Nasco
Supplier: ORS Nasco
Rollgliss™ Rollgliss®
098-8902004
300058-336EA 5526.46 USD InStock
300058-336
Rollgliss® Rope Rescue Systems, ORS Nasco
Fall Protection Fall Protection Kits
  • 50 feet of travel distance
  • Other lengths and ratios available
  • This pre-rigged/pre-engineered rope rescue system provides 3:1 rescue ratio
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty