Would you like to log in again?
Industrial Heat Guns, Weller®
Supplier: Weller®
185-6966C
300062-980EA 370.13 USD InStock
300062-980
Industrial Heat Guns, Weller®
Heaters Hot Air Dryers
Designed for assembly and repair applications in production plants, laboratories and service shops.
  • Long-life heater and fan components
  • 3-position switch Hot-Off-Cool
  • 3-wire grounded plug 6' power cord
  • For epoxy curing, drying, cooling, and shrinking tubing and vinyl/T.F.E
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty