Would you like to log in again?
'PT' Series Soldering Iron Tips, Weller®, ORS Nasco
Supplier: ORS Nasco
185-PTA7
300063-096EA 10.27 USD InStock
300063-096
'PT' Series Soldering Iron Tips, Weller®, ORS Nasco
Soldering Supplies Soldering Irons
  • Precision ground
  • Provides maximum heat transfer from heater to tip
  • IMPORTANT: Use only Weller® tips. Use of non-Weller® component causes damage to the unit
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty