Would you like to log in again?
Analog Soldering Stations, Weller®, ORS Nasco
Supplier: ORS Nasco
185-WES51
300063-152EA 308.09 USD InStock
300063-152
Analog Soldering Stations, Weller®, ORS Nasco
Soldering Supplies Soldering Stations
  • New receptacle for easier iron replacement
  • Designed for continuous production soldering
  • Slim, comfortable pencil, with ETA tip, reduces operator fatigue
  • Tip temperature offset capability allows user to reset station temperature to match in tip sizes & styles
  • Iron is fitted with a non-burnable silicon rubber cord for safety
  • ESD safe to protect sensitive components
  • Use ET series tips
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty