Would you like to log in again?
High Temp 44 Markers, Dykem, ORS Nasco
Supplier: ORS Nasco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
253-44424 253-44250 253-44219
300065-738PK 202.12 USD InStock
300065-738 300065-736 300065-744
High Temp 44 Markers, Dykem, ORS Nasco
Pens Permanent Markers
High temperature marker for use on glass, most metals or ceramic.
  • Medium size fiber tip
  • Pigmented and oil-based
  • Pen cap clip fits on pocket or clipboard
  • Marks become permanent and fused when heated
Full cure time: 1 min at 77 °F
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty