Would you like to log in again?
DYKEM® BRITE-MARK® Medium Markers, ORS Nasco
Supplier: ORS Nasco
DYKEM® BRITE-MARK®
253-84009 253-84001
300000-770PK 61.36 USD InStock
300000-770 300065-772
DYKEM® BRITE-MARK® Medium Markers, ORS Nasco
Pens Permanent Markers
  • Acrylic paint makes permanent marks on most surfaces, dries quickly
  • Valve action mechanism prevents paint from drying in barrel
  • Heavy duty barrel
  • Bullet tips are replaceable
  • Reistant to acids, alkalis, oil, gasoline, water, and weather
Acrylic paint makes permanent bright marks on metal, rubber, plastic, glass, concrete, leather, stone, wood, vinyl, and more. Ideal for color coding. Valve action mechanism prevents paint in barrel from drying when cap is left off. Heavy duty barrel retains its shape. Bullet tips are replaceable to assure complete use of paint. Resistant to acid, alkali, oil, gasoline, petroleum, water, and weather. Drying time: 25–35 seconds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty