Would you like to log in again?
Regulator Outlet Bushings, Western Enterprises
Supplier: Western Enterprises
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
312-D-33 312-137 312-132 312-C-34 312-135 312-144 312-C-40 312-33 312-D-34 312-142 312-133 312-C-35 312-134 312-145 312-C-32 312-32 312-D-32 312-136 312-C-33 312-143
300066-764EA 36.42 USD InStock
300066-764 300066-236 300066-766 300066-226 300066-232 300007-837 300066-242 300066-696 300066-762 300066-690 300066-250 300066-692 300066-252 300007-951 300066-240 300007-842 300007-952 300007-796 300007-794 300007-792
Regulator Outlet Bushings, Western Enterprises
Welding Supplies
  • Made from the highest quality material to ensure functionality and durability
  • Used to reduce an outlet in a fitting or to connect a pipe to a larger outlet
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty