Would you like to log in again?
Manifold One-Piece Dust Cap Assemblies, Western Enterprises
Supplier: Western Enterprises
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
312-DC-510 312-DC-B7 312-DC-B8 312-DC-580
300066-780EA 13.12 USD InStock
300066-780 300066-778 300066-782 300066-784
Manifold One-Piece Dust Cap Assemblies, Western Enterprises
Welding Supplies
One-Piece Dust Cap Assemblies are ideal for dust control in welding applications.
  • Made from the highest quality material to ensure functionality and durability
  • Can accommodate up to 200 psi of pressure
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty