Would you like to log in again?
Regulator Gauges, Western Enterprises
Supplier: Western Enterprises
312-G-18-4000W 312-G-2-400W 312-G-2-4000W 312-G-2-30RLW 312-G-25-200W 312-G-2-200W 312-G-2-30W 312-G-25-400W
300066-826EA 47.9 USD InStock
300066-826 300066-828 300007-983 300066-822 300066-824 300066-830 300066-832 300066-820
Regulator Gauges, Western Enterprises
Manometers
Installation requires PTFE tape or oxygen safe compound to guard against leakage
All gauges feature a 1/4" (NPT) thread size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty