Would you like to log in again?
RM Single Stage Manifold Regulators, Western Enterprises
Supplier: Western Enterprises
312-RM-4-4 312-RM-7-4
300066-950EA 620.53 USD InStock
300066-950 300066-952
RM Single Stage Manifold Regulators, Western Enterprises
Welding Supplies
Pressure compensated single stage design delivers stable delivery pressure performance.
Unique cartridge permits easy, single-unit replacement of vital parts in minutes. This includes easy replacement of the seat, nozzle, filter, spring, seals, and built-in check-relief valve. Cartridge can easily be removed and installed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty