Would you like to log in again?
Pedestal Mounted Eye Washes, Guardian
Supplier: Guardian
333-G1825BC
300068-322EA 924.63 USD InStock
300068-322
Pedestal Mounted Eye Washes, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Free standing, pedestal mounted eye wash, water flow is activated by either flag handle or foot treadle.
  • Valve: ¹/₂" IPS chrome plated brass stay-open ball valve, is US-made with chrome plated brass ball and Teflon™ seals
Pipe and fittings: schedule 40 galvanized steel, furnished with orange polyethylene covers for vertical piping for high visibility and corrosion resistance.
Two GS-Plus™ spray heads deliver flood of water for rinsing eyes, each head has a 'flip top' dust cover, internal flow control and filter to remove impurities from the water flow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty