Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eye Wash & Shower Stations, Guardian

Supplier: Guardian

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

333-G1902 333-G1902HFC 333-G1902P
300068-326EA 1272.07 USD InStock
300068-326 300068-328 300068-330
Eye Wash & Shower Stations, Guardian
First Aid Supplies Eyewash Stations and Eyewash Bottles
Eye wash is activated by flag handle, two GS-Plus™ spray-type outlet heads deliver water for rinsing eyes.

  • Eye Wash Valve: 1/2" IPS chrome-plated brass stay-open ball valve
  • Shower Valve: 1" IPS chrome-plated brass stay-open ball valve, furnished with stainless steel actuating arm and 29" stainless steel pull rod.
  • Spray head assembly: two GS-Plus™ spray heads each have 'flip top' dust cover, internal flow control and filter to remove impurities from the water flow.
  • Pipe and fittings: schedule 40 galvanized steel, furnished with orange polyethylene covers for vertical piping for high visibility and corrosion resistance.

Bowl height: 36", bowl dia. 11.5" or 12"
Outlet: 1.25"
Shower head Safety Orange ABS dia. 10x85"

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty