Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Series 600 Trucks, Harper Trucks

Supplier: Harper Trucks

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

338-60072-64 338-650A50-51 338-65050-51 338-60068-77 338-65029-51
300068-612EA 1561.74 USD InStock
300068-612 300068-614 300068-616 300008-179 300068-610
Series 600 Trucks, Harper Trucks
Carts
Available in two and four wheel models and is designed to handle heavy CRYOGENIC LIQUID GAS CYLINDERS.

  • This larger sized frame and repositioned front wheels which allow secure handling without excessive force
  • The 650A50-51 is designed to handle larger 20" -26" diameter liquid gas cylinders
  • This model equipped with the QUAD Hook will handle a wide variety of large gas cylinders
  • Newly designed single hook adjusts from 50" to 60" in height which makes it more versatile for handling a variety of cylinder types
  • All available with pneumatic or mold on rubber tires

Colour: Harper Green

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty