Would you like to log in again?
Advantage® Respirator Cartridges, MSA
Supplier: MSA
Advantage®
454-815369 454-815359 454-815368 454-815366 454-815362 454-815355
300072-652PK 65.86 USD InStock
300072-652 300072-650 300072-646 300009-019 300009-020 300072-648
Advantage® Respirator Cartridges, MSA
Respirators Reusable Respirators Respirator Filters
Advantage® line of particulate, chemical, and combination cartridges are lightweight, low-profile performance.
  • Advantage® line of particulate, chemical, and combination cartridges are lightweight, low profile performance
  • Cartridges fit both 200 LS Half mask and 3000 Full-facepiece advantage respirators
  • Self-adjusting crown straps
  • Consists of a polyethylene shell and suspension system working together as a protection system
  • Also fits 420 Half-mask respirators and 1000, 3000, and 4000 Full-mask respirators
Certifications: NIOSH Approved to 42 CFR Part 84.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty