Would you like to log in again?
N-Ferno® 6990 Hand-Style Warming Packs, Ergodyne
Supplier: ORS Nasco
N-Ferno®
150-16990
300073-624CS 181.91 USD InStock
300073-624
N-Ferno® 6990 Hand-Style Warming Packs, Ergodyne
First Aid Supplies First Aid Kit Components
Hand style warming packs warms up to 8 hours.
  • All natural ingredients heat up when they contact oxygen
  • Seal from oxygen in Zip-Loc bag and reuse the next day
  • Self-contained packs add heat anywhere when exposed to air
  • To activate, just open the pack
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty