Would you like to log in again?
EZ Stop® III Shock Absorbing Lanyards, ORS Nasco
Supplier: ORS Nasco
EZ Stop®
098-1244006
300075-590EA 151.6 USD InStock
300075-590
EZ Stop® III Shock Absorbing Lanyards, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Limits arresting forces to 900 lbs. or less
  • Soft cover shock absorber for comfort
  • 1-3/8" tubular web jacket and polyester core for durability and longevity
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty