Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Concrete Detent Anchors, DBI-SALA, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-2101004
300075-608EA 494.26 USD InStock
300075-608
Concrete Detent Anchors, DBI-SALA, ORS Nasco
Fall Protection Fall Arresters
It allows anchor to be moved from location to location.
  • Anchor receptacle is installed in concrete
  • Good for permanent installs where visual appearance needs to be pleasing
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty