Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Protecta Cable Sling Tie Off Adaptors, ORS Nasco
Supplier: ORS Nasco
098-AJ408AG
300075-724EA 98.22 USD InStock
300075-724
Protecta Cable Sling Tie Off Adaptors, ORS Nasco
Fall Protection Fall Arresters
This temporary anchorage connector/anchorage ring easily accommodates various snaphooks and carabiners.
  • Coating protects anchor point and wire rope
  • Versatile, fits a variety of anchorage points
  • Includes 6' cable
Certifications: Meets OSHA 1926 and ANSI Z359 Standards.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty