Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

440 Basic-DV™ Direct Vent Goggles, Bouton®

Supplier: Bouton
Basic-DV™
112-4400-400 112-4400-300
300051-864EA 5.2 USD InStock
300051-864 300076-134
440 Basic-DV™ Direct Vent Goggles, Bouton®
Goggles
Lenses made of polycarbonate block 100% of harmful UV Radiation.

  • Made in the USA
  • With elastic strap attachment

Weight: 0.08 lbs./0.15 lbs.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty