Would you like to log in again?
Cordless XRP™ Li-Ion Impact Drivers, DeWalt®, ORS Nasco
Supplier: ORS Nasco
Cooper Center™
115-DC827KL
300076-732EA 704.23 USD InStock
300076-732
Cordless XRP™ Li-Ion Impact Drivers, DeWalt®, ORS Nasco
Wrenches
This tool allows access into tighter areas and reduces user fatigue.
  • Frameless motor with replaceable brushes for extended tool durability and life
  • Textured ant-slip comfort grip provides maximum comfort
  • Drive type: hex shank quick release
Delivery information: Supplied incl. 1 hour charger, heavy-duty kit box, (2) 18V Li-Ion batteries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty