Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Stepless® Ear Clamp Kits, Oetiker
Supplier: Oetiker
Stepless®
320-18500060
300078-906EA 102.58 USD InStock
300078-906
Stepless® Ear Clamp Kits, Oetiker
Clamps Tubing and Sanitary Clamps
These clamps provide a smooth inner band and uniform compression for a 360° seal.
  • Kits contain assorted sizes of stainless steel Stepless® Ear Clamps
  • Suitable for demanding applications where lighter, thin-walled hose or tubing is being used
Ordering information: Kits include (1) Standard Jaw Pincers, (10) of 11/4", (10) of 11/8", (10) of 1", (10) of 1/2", (10) of 23/32", (10) of 5/8", (10) of 7/8", (10) of 9/16".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty