Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Battery Operated Smoke Alarms, KIDDE, ORS Nasco

Supplier: ORS Nasco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

408-PE9E
300080-128EA 52.81 USD InStock
300080-128
Battery Operated Smoke Alarms, KIDDE, ORS Nasco
Smoke Detectors
This smoke alarm features a red LED indicator, which flashes every 30-40 seconds to indicate that the alarm is receiving power

  • With test button and low-battery indicator
  • Temperature range 4.4 to 37.8 °C (40 to 100 °F)
  • 85 dB sound level
  • For use with 9 V battery

Available with dust cover.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty