Would you like to log in again?
Advantage® 420 Series Reusable Half Mask Respirators, MSA
Supplier: MSA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Advantage®
454-10102182 454-10102184
300081-010EA 48.69 USD InStock
300081-010 300081-014
Advantage® 420 Series Reusable Half Mask Respirators, MSA
Respirators Reusable Respirators Half Face Respirators
With its UniBond over-mold facepiece, AnthroCurve™ face seal, and low profile design, the Advantage® 420 Respirator is the superior option for respiratory protection.
The over-mold facepiece and face seal ensure the unit provides an excellent fit and comfort to the wearer. The unique yoke and harness design (patent pending) allows a user to easily don and doff the state-of-the-art respirator while maintaining a customized fit. Other features include a ComforTop headband, a built-in moisture release chin cup hole, and the extra-wide head and neck straps.
Delivery information: Respirators are supplied with facepiece, head cradle, elastic straps, buckle set, yoke withe lever, (2) inhalation valves, and (1) exhalation valve.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty