Would you like to log in again?
M12™ Compact Copper Tubing Cutters, Milwaukee Electric Tools
Supplier: Milwaukee Electric Tools
M12™
495-2471-21
300081-918EA 421.62 USD InStock
300081-918
M12™ Compact Copper Tubing Cutters, Milwaukee Electric Tools
Cutters
M12™ compact copper tubing cutter are designed to cut the tubing of sizes from 1/2" to 11/8" dia.
  • Built-in LED light
  • All metal sealed front end
  • Auto adjusting jaws
  • Electronic variable control type
  • Ultra-efficient drive mechanism
The auto-locking cut mechanism of this cutter provides 200 cuts per charge.
Delivery information: Supplied with lithium-ion battery (1.400 A-h), 30 minute charger and carrying case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty