Would you like to log in again?
Vortec Pro® Type 1 Grinding Wheels, Weiler®
Supplier: Weiler
Vortec Pro®
804-36459 804-36460 804-33053 804-33052
300085-638CS 20 USD InStock
300085-638 300085-640 300085-388 300085-390
Vortec Pro® Type 1 Grinding Wheels, Weiler®
Abrasives
  • For use on right angle grinders, electric or air die grinders, tool room grinders and circular saws
  • Designed for general purpose grinding and cutting applications on steel, iron, and other ferrous metals
  • Wheels are resin-bond formulated for aggressive stock removal and fast cutting, and are reinforced with fiberglass webbing and a metal ring around the hub for additional strength and stability
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty