Would you like to log in again?
Weiler® Vortec Pro Stem Mounted Crimped Wire End Brush, ORS Nasco
Supplier: Weiler
804-36281 804-36248 804-36285 804-36046 804-36047 804-36247 804-36284 804-36048 804-36049 804-36280 804-36249 804-36246 804-36282 804-36283
300085-606CS 156.42 USD InStock
300085-606 300085-598 300085-476 300085-564 300085-600 300085-478 300085-566 300085-602 300085-568 300085-604 300085-480 300085-482 300085-570 300085-596
Weiler® Vortec Pro Stem Mounted Crimped Wire End Brush, ORS Nasco
Brushes
High impact action for severe applications in tight areas.
  • Brushes can also be used for rubber and plastic removal
  • Remove large burrs and heavy contamination
Applications include paint removal, deburring, and cleaning and finishing recessed areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty