Would you like to log in again?
Master-Mite® Heat Guns, Master Appliance
Supplier: Master Appliance
Ecoheat™
467-10008
300087-858EA 313.99 USD InStock
300087-858
Master-Mite® Heat Guns, Master Appliance
Heaters Hot Air Dryers
It is used to activate adhesive, cure epoxies, dry components, melt solder preforms, remove decals, shrink tubing and more.
  • Complete system comes standard with 650 °F heating element, adjustable stand and heat shrink attachment
  • Performs benchtop heating jobs without excess air flow, won't blow components around work station
  • Quiet running motor, eliminates noisy production areas
  • Made in USA
Fan speed: Single
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty