Would you like to log in again?
Non-Corrosive Gas Flow Valves, Western Enterprises
Supplier: Western Enterprises
312-208-F 312-218-D 312-210 312-218 312-217 312-215-FR 312-206 312-207 312-205 312-203 312-211 312-215-AR 312-204 312-211-E 312-208
300007-818EA 37.48 USD InStock
300007-818 300007-816 300007-817 300066-298 300087-802 300007-814 300007-815 300066-286 300066-288 300007-813 300066-296 300091-790 300066-284 300066-292 300091-794
Non-Corrosive Gas Flow Valves, Western Enterprises
Welding Supplies
  • Made from brass to ensure functionality and durability
  • Use these reliable non corrosive gas flow valves with all non-corrosive gases including oxygen
  • Can withstand elevated pressures
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty