Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Tiger Paw Coated Abrasive Flap Discs, Weiler®
Supplier: Weiler
804-51107 804-51115 804-51109 804-51114 804-51112 804-51110 804-51108 804-51113
300053-342CS 103.77 USD InStock
300053-342 300053-340 300053-338 300053-336 300053-348 300092-028 300053-346 300053-344
Tiger Paw Coated Abrasive Flap Discs, Weiler®
Abrasives
Multiple layers of coated abrasive material expose sharp, fresh grains as they slowly wear away, and the 100% premium zirconium grain prevents premature grain shedding on aggressive applications resulting in increased productivity and reduced operating costs.
  • Designed for aggressive performance on tough grinding applications
  • High performance grain and special poly cotton cloth provide superior grain retention
  • This combination gets the job done faster and reduces overall grinding and finishing costs
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty