Would you like to log in again?
Proto® Standard Length Round Head Ratchets, ORS Nasco
Supplier: ORS Nasco
577-5452F
300092-854EA 118.4 USD InStock
300092-854
Proto® Standard Length Round Head Ratchets, ORS Nasco
Tool Sets
Small head design and tight swing allow fastener to access tight areas.
  • 5° ratcheting angle
  • 1.5" head width
  • Full polish shaft with knurled grip for strong slip-resistant grip
Certifications: ASME B107.10M.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty