Would you like to log in again?
Phlebotomy Tray Units, JAC Medical
Supplier: JAC Medical Products
JAC10 JAC59 JAC9 JAC59 JAC8 JAC8 JAC57 JAC9 JAC58 JAC10 JAC58 JAC60 JAC60
30109-518EA 196.56 USD InStock
CA78010-016 30109-518 CA78010-148 CA78010-018 62687-306 CA78010-020 30109-522 30109-502 30109-524 CA78010-144 CA78010-146 30109-520 62687-300
Phlebotomy Tray Units, JAC Medical
Trays/Dishes/Pots Blood Collection Trays
Polystyrene trays include components to assist in efficient blood collection
All replacement components (except HDPE containers) are constructed of translucent white autoclavable polypropylene. The utility boxes contain one removable divider and are useful for carrying a variety of small items. The gauze box has one removable divider that creates three compartments total: one large 11.7Wx11.7Dcm (45/8x45/8") compartment and two smaller 6.4Wx7Dcm (21/2x23/4") compartments. The packet/tube holder is ideal for BD Vacutainer® tubes, packets, winged infusion sets, and syringes. It features three removable dividers to provide up to six compartments. The 237mL (8oz.) HDPE containers feature snap-on lids and have excellent resistance to acids, alcohols, and bases.
Ordering information: Mini tray unit 30109-522 is supplied with one 11-well test tube rack, two mini utility boxes, one small waste holder, and two 237mL (8oz.) HDPE containers with lids. Medium tray units 30109-520 and 62687-300 are supplied with a test tube rack, two utility boxes, one small waste holder, two 237mL (8oz.) HDPE containers with lids, one slip holder, and one slide holder.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty