Would you like to log in again?
SensaTemp® Handpieces, PACE®
Supplier: Pace
SensaTemp®
6993-0153-P1 1121-0330-P1 1121-0316-P1 7023-0002-P1 7025-0001-P1 1121-0365-P1 1121-0352-P1 1121-0483-P1 1121-0382-P5 1121-0317-P1 1121-0318-P1 7024-0001-P1 1121-0320-P1 1121-0456-P1 11210322002 1121-0321-P1 1121-0484-P1 1121-0324-P1 1121-0383-P5 1121-0326-P1 1121-0351-P1 1121-0323-P1 11210322001 1121-0325-P1 1121-0319-P1
30138-078EA 148.97 USD InStock
30138-078 30138-110 30138-074 30138-194 30138-076 30138-130 30138-070 30138-094 30138-072 30138-192 30138-090 30138-108 30138-128 30138-122 30138-364 30138-088 30138-366 30138-068 30138-360 30138-184 30138-084 30138-362 30138-086 30138-080 30138-082
SensaTemp® Handpieces, PACE®
Soldering Supplies Desoldering Tools
For use with PACE® SensaTemp® compatible stations
SensaTemp® handpieces have a tip temperature range of 204 to 482°C (400 to 900°F) with a temperature stability of ±1.1°C (2°F) at idle from set tip temperature. Choose from the TP-65 ThermoPik™ SMD removal handpiece with vacuum pick-up, the TJ-70 ThermoJet® Hot Air SMD handpiece, and the TT-65 ThermoTweez® SMD removal handpiece.
Ordering information: Handpieces and tips sold separately. Vacuum cup kit 30138-360 includes one small, one medium, and one large cup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty