Would you like to log in again?
SX-80 SensaTemp® Sodr-X-Tractor™ Desoldering Handpiece, PACE®
Supplier: Pace
SensaTemp® Sodr-X-Tractor™
1309-0054P10 1121-0681-P5 1121-0682-P5 1121-0628-P5 1121-0629-P5 1121-0624-P5 1121-0678-P5 1121-0627-P5 1121-0630-P5 1121-0631-P5 1121-0679-P5 1121-0626-P5 1121-0625-P5 6010-0106-P1 1121-0680-P5 1121-0690-P5
30138-254PK 109.33 USD InStock
30138-254 30138-264 30138-242 30138-352 30138-256 30138-244 30138-250 30138-260 30138-252 30138-262 30138-240 30138-258 30138-246 30138-268 30138-348 30138-248
SX-80 SensaTemp® Sodr-X-Tractor™ Desoldering Handpiece, PACE®
Soldering Supplies Desoldering Tools
Features an improved design with a disposable solder collection chamber and quick-change heating element
Handpiece uses Endura™ desoldering tips. These nonclogging tips require less maintenance and cleaning.
Thru-hole desoldering, thermo-drive desoldering, flo desoldering, PCMCIA fine pitch TSOP removal, and small PQFP removal are possible with this unit. For use with SensaTemp® stations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty