Would you like to log in again?
Nalgene® Analytical Funnels, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
4250-0055 4250-0160 4250-0100 4250-0065 4250-0075 4250-0035
30253-140CS 396.55 USD InStock
30253-140 30253-060 30253-082 30253-129 30253-049 30253-027
Nalgene® Analytical Funnels, Polypropylene, Thermo Scientific
Funnels Filtration Funnels Analytical Funnels
Funnels are designed for analytical chemistry.
Accept standard size filter paper. Body of funnel makes exact 60° angle. Internal ribs and 58° angle for rapid filtration. Outside ribbing prevents airlock. Autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty