Would you like to log in again?
Whatman™ Polycap™ TC Disposable Filter Capsules
Supplier: Whatman products (Cytiva)
Whatman™ Polycap™ Polycap™
6714-7501 6717-9502 6717-7510 6715-3601 6704-9502 6718-9582 6714-7501 6715-3682 6717-7510 6717-9504 6715-3602 6717-9501 6716-3602 6714-3601 6715-3682 6717-9501 6704-9502 6715-3604 6715-7502 6717-7504 6717-7504 6718-9502 6714-7502 6715-3602 6717-9502 6714-3602 6715-7582 6714-3601 6716-3602 6718-9502 6717-9504 6714-3602 6715-7582 6714-3604 6715-7502 6715-3601 6714-7502 6718-9582 6715-3604 6714-3604
83009-778EA 105.32 USD InStock
CA11013-756 CA83009-780 CA83009-782 CA30253-228 83009-778 30253-242 30253-240 30253-262 30253-260 30253-228 CA10147-954 CA30253-258 30253-248 30253-226 30253-224 CA30253-234 30253-244 30253-222 CA30253-232 83009-780 83009-782 CA11012-562 CA83009-778 CA30253-240 CA30253-262 30253-232 10035-484 CA30253-260 CA30253-248 CA30253-226 CA11013-916 10147-954 CA30253-224 10035-486 CA30253-222 30253-258 CA30253-244 10035-488 30253-234 CA30253-242
Whatman™ Polycap™ TC Disposable Filter Capsules
Filters Capsule Filters
Whatman Polycap TC filtration capsules from Cytiva feature dual layer polyethersulfone (PES) membranes and provide efficient filtration for aqueous solutions.
  • 100% integrity-tested during manufacturing
  • Polypropylene housing thermally fused to eliminate surfactants and mold releasing agents that could affect analysis
  • Available in sterile and nonsterile versions with a filling bell option
  • PES membranes protein adsorption characteristics: HAS 0.4 µg/cm²; Insulin 2.0 µg/cm²; gammaglobulin 1.5 µg/cm²
PES membranes are inherently hydrophilic, have low extractables, are biosafe, and have excellent flow rates. These characteristics make Polycap TC PES filters appropriate for a variety of critical filtration applications, including cleaning/rinsing pharmaceutical solutions, tissue culture media, and viral suspensions.
Certifications: Manufactured according to ISO9001:2008 QMS standards.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty