Would you like to log in again?
Industrial Funnel, Polyethylene, Dynalon
Supplier: Dynalon
342345
30286-503CS 482.3 USD InStock
30286-503
Industrial Funnel, Polyethylene, Dynalon
Funnels Filtration Funnels Filter Funnels
Large industrial funnels made of polyethylene
Designed for transferring liquids. Maximum temperature for HDPE is 110°C
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty