Would you like to log in again?
Chemware® Funnel, Teflon® PFA, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
Chemware®
D1069642
30286-600EA 289.46 USD InStock
30286-600
Chemware® Funnel, Teflon® PFA, Saint-Gobain Performance Plastics
Funnels Filtration Funnels Filter Funnels
For ultra-pure liquid transfer
Teflon® PFA is non-contaminating and inert. Low surface energy facilitates quantitative transfer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty