Would you like to log in again?
PYREX® Separatory Funnels, Squibb, Pear-Shape, Fluoropolymer Resin Stopcock, Corning
Supplier: Corning
PYREX®
6404-125 6404-1L 6404-2L 6404-60 6404-250 6404-500
89091-508CS 667.99 USD InStock
89091-508 30356-700 30356-722 89090-654 89090-792 89090-880
PYREX® Separatory Funnels, Squibb, Pear-Shape, Fluoropolymer Resin Stopcock, Corning
Funnels Separating Funnels
PYREX® Borosilicate glass.
With PTFE stopcock and linear, high-density, closed-bottom polyethylene stoppers which conform to [ST] stopper dimensions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty