Would you like to log in again?
SP Bel-Art Separatory Funnel and Imhoff Cone Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SCIENCEWARE®
H18967-0000
30534-905EA 485.49 USD InStock
30534-905
SP Bel-Art Separatory Funnel and Imhoff Cone Rack, Bel-Art Products, a part of SP
Support Stands
This high-density polyethylene rack holds four separatory funnels on the lower shelf or four long-stem funnels on the upper shelf (maximum funnel diameter is 98 mm).
Set screws allow shelf height adjustments anywhere along the 74 cm support rods.
Delivery information: Rack includes 24 insert clips that allow the rack to accommodate different sized separatory funnels and imhoff cones.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty