Would you like to log in again?
SP Bel-Art Write-On™ Label Tape, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Write-On™
F13480-0075 F13490-0075 F13483-0050 F13490-0050 F13486-0075 F13484-0050 F13488-0050 F13482-0100 F13485-0100 F13482-0075 F13485-0050 F13486-0100 F13488-0075 F13486-0050 F13487-0050 F13481-0100 F13487-0100 F13483-0100 F13480-0100 F13462-0300 F13481-0075 F13490-0100 F13482-0050 F13463-0600 F13480-0050 F13483-0075 F13487-0075 F13481-0050 F13488-0100 F13484-0075 F13484-0100 F13485-0075
30617-014CS 139.08 USD InStock
30617-014 30617-036 30617-058 30617-016 30617-038 30617-054 30617-032 30617-010 30617-012 30617-034 30617-056 30617-018 30617-060 30617-062 89005-240 30617-040 30617-024 30617-046 30617-026 30617-048 30617-042 30617-020 30617-064 30617-022 30617-044 30617-028 30617-006 30617-008 89005-238 30617-050 30617-052 30617-030
SP Bel-Art Write-On™ Label Tape, Bel-Art Products, a part of SP
Tapes
Adheres to any clean surface, including Teflon® fluoropolymer resin-coated materials, and peels off without leaving a sticky residue.
  • Resists moisture and most solvents
  • Withstands autoclaving and gamma irradiation
  • Suited for use in low-temperature applications
Pencil, ball-point pen, or solvent ink marker remain legible under the most demanding laboratory conditions
The tape will withstand a maximum temperature of 157°C (315°F) for 30 minutes or 80°C (176°F) for 24 hours. It can be used in temperatures as low as –80°C (–112°F). Tape will withstand a steam autoclaving cycle at 121°C (250°F) and gamma irradiation between 13.0 and 45.0kGy.
Consists of colored paper with rubber resin adhesive that resists moisture and most solvents. Available in six vivid colors and great for use in low-temperature applications.
Ordering information: Excellent for use with Write-On™ Multi-Roll Label Tape Dispensers (not included). Assorted color pack 89005-238 includes one roll each of green, yellow, and white. Assorted color pack 89005-240 includes one roll each of blue, green, orange, red, white, and yellow. Cover with Scienceware® Clear Protective Tape for extreme temperature and chemical protection. Tape rolls with 2.5 cm (1") dia. core fit Scienceware® dispensers 74950-074 and 74950-072. Tape rolls with 7.6 cm (3") dia. core fit Scienceware® dispenser 36443-003, 74950-074 and 74950-072.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty