Would you like to log in again?
White Polypropylene Containers, Dynalon
Supplier: Dynalon
426325-0400 426325-1600
30617-166CS 226.72 USD InStock
30617-166 71070-188
White Polypropylene Containers, Dynalon
Sample Containers General Purpose Containers
Containers are molded from white opaque polypropylene and are not considered leak-proof
They provide many laboratory and industrial uses. Supplied with polypropylene screw closures. Maximum operating temperature for polypropylene is 121°C (250°F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty