Would you like to log in again?
Bottle, Wide Mouth, HDPE, Dynalon
Supplier: Dynalon
606215-0016
30617-172CS 2019.57 USD InStock
30617-172
Bottle, Wide Mouth, HDPE, Dynalon
Bottles Plastic Bottles
These general-purpose HDPE bottles are leak-proof, and very economical.
  • Closure is made of LDPE
Maximum operating temperature for HDPE is 110 °C (230 °F), and 80 °C (176 °F) for LDPE.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty